Cole-Parmer Electrode pH Soil Glass BNC
Representative image only.
Cole-Parmer Item # RK-05992-62
Soil measurements without the "Suspension effect"
  • Refillable, glass-body electrode allows you to immerse the bulb without immersing the ceramic junction
  • 6 month warranty
$332.99CAD / Each
BUY MORE & SAVE
  • 1-3$332.99CAD / Each
  • 4+$299.69CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeSingle junction
  • Electrode Body MaterialGlass
  • Reference ElectrolyteAg/AgCl
  • Max Temperature (° C)100
  • ConnectorBNC
  • DescriptionELECTRODE PH SOIL GLASS BNC
  • Warranty6 month warranty
6 Month Warranty
Key Features
  • Refillable, glass-body electrode allows you to immerse the bulb without immersing the ceramic junction
  • 6 month warranty
More About this Item
Our refillable soil electrode has an extra-wide 7.5 mm ceramic junction. Soil electrodes have a maximum temperature range of 212°F
(100°C) and measure 145 mm L x 12 mm dia. Ag/AgCl ref cell.
Use with any meter that accepts a BNC connector, or check out our selection of pH/Ion meters.