Cole-Parmer E1101 Mini Electrophoresis System; 115 V, 50-60 Hz

Representative image only.
Cole-Parmer Item # RK-28559-11
Quick and easy, no spacers or clamps required
  • Visualize your run with the viewing area above the gel box
  • Quickly run gels with selectable parameters using the membrane keypad
  • Compact footprint ideal for small laboratories or classrooms
  • 3 year warranty

PRODUCT OPTIONS

$987.17CAD / Each
Include Accessories Today!

SPECS & DESCRIPTION

Specifications & Description

  • Buffer Solution Required (mL)230
  • Gel CapacityOne 10.5 x 6 cm or two 5 x 6 cm
  • Glass Width (in)4.3
  • Glass Length (in)4.7
  • Glass Height (in)1.8
  • Glass Width (cm)11
  • Glass Length (cm)12
  • Glass Height (cm)4.5
  • Length (in)7 1/2
  • Height (in)2 1/5
  • Width (in)5 1/10
  • Length (cm)19
  • Height (cm)5.5
  • Width (cm)13
  • Power (VAC)115
  • DescriptionMini electrophoresis system; 115V
  • Warranty3 Years
3 Year Warranty

Includes

US plug.

Key Features

  • Visualize your run with the viewing area above the gel box
  • Quickly run gels with selectable parameters using the membrane keypad
  • Compact footprint ideal for small laboratories or classrooms
  • 3 year warranty

More About this Item

This complete horizontal mini electrophoresis system includes everything needed to get up and running. The system is designed for separating nucleic acids in agarose gel. Selectable parameters include three preset voltages – 35, 50, and 100V, as well as a timer that has a range of 0 to 99 minutes or for continuous use. The safety lid is vented to dissipate heat during the electrophoresis process. The current will only run through the system when the lid is properly in place.
The gel trays feature a dark contrast strip to help visualize wells while loading. In addition, the lines fluoresce under UV and blue light to help track the progress of the sample during the run.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat