Cole-Parmer Essentials Graduated Cylinder w/Certificate of Graduation Accuracy, NIST Calibration; 100 mL

Representative image only.
Cole-Parmer Essentials Item # RK-34300-24
Feel more confident about your aliquots - meet ISO 9000 requirements
  • Single metric scale
  • Class A cylinder glass meets ASTM E 438
  • Autoclavable

PRODUCT OPTIONS

$302.04CAD / Each

$131.87 - $238.69CAD / Each

Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • MaterialGlass
  • Capacity (mL)100
  • Graduation Interval (mL)1
  • Tolerance (mL)±0.40
  • DescriptionGraduated Cylinder w/Certificate of Graduation Accuracy, NIST Calibration; 100 mL

Key Features

  • Single metric scale
  • Class A cylinder glass meets ASTM E 438
  • Autoclavable

More About this Item

These glass graduated cylinders feature an easy-to-read white scale and a molded base to decrease risk of accidentally knocking it over.

All support ISO 9000 requirements and are calibrated in accordance with ASTM E 542. Cylinders meet ASTM E 694 specifications and accuracy requirements of ASTM 1272. The calibration is traceable to a National Metrology Institute recognized by the Bureau International des Poids et Mesures which could include NIST (USA), CENAM (México) among others.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat