Nelson-Jameson Chloride Analyzer; 120/220 VAC, 50/60 Hz

Representative image only.
Item # RK-02656-20
More accurate and precise than any other rapid method of salt determination!
  • Measurement repeatability ensures quality assurance and control
  • Compact design saves valuable bench space
  • Chloride standards are calibrated in both mg/L and mmol/L
  • 1 year warranty

$18,661.96CAD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Accessory TypeChloride Analyzer
  • Range10 to 999 mg/L chloride, 2 to 165 mg% salt
  • ReproducibilityCV < 1% for 20 replicate samples at 200 mg/L
  • Resolution1 mg/L chloride
  • Accuracy±3 mg/L at the 200 mg/L level
  • OutputRS-232
  • Power (VAC)120 / 220
  • Length (in)9.75
  • Width (in)7.75
  • Height (in)12.5
  • Length (cm)24.8
  • Width (cm)19.7
  • Height (cm)31.8
  • DescriptionChloride Analyzer; 120/220 VAC, 50/60 Hz
  • Warranty1 year
1 Year Warranty

Key Features

  • Measurement repeatability ensures quality assurance and control
  • Compact design saves valuable bench space
  • Chloride standards are calibrated in both mg/L and mmol/L
  • 1 year warranty

More About this Item

This automatic chloride titrator provides accurate and highly reproducible measurement results. The system is ideal for use in the food, pharmaceutical, agricultural, and petroleum industries. The system utilizes a method based on coulometric titration — an absolute method where the reagent, silver ions, is precisely and quantitatively generated at the time of analysis by passing a constant current between electrodes. The end point is detected by the use of sensing electrodes which measure the change in the solution conductivity that occurs when excess silver ions are present in the solution. Unlike a conductivity meter, this analyzer actually titrates chloride with silver as in official methods.
Automatic features provide speed and simplicity, reducing labor costs and

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat