Cole-Parmer Essentials Bottletop Dispenser, Recirculation Valve, 10 to 100 ML

Representative image only.
Cole-Parmer Essentials Item # RK-25208-37
Save money by conserving reagents
  • Recirculation valve prevents loss of reagent during priming and purging
  • Smooth plunger movement provides bubble-free dispensing
  • Adjustable nozzle allows you to change the dispensing angle
  • 1 year warranty

PRODUCT OPTIONS

$1,156.61CAD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Min Volume (mL)10
  • Max Volume (mL)100
  • Division (mL)2
  • Accuracy±0.6%
  • Reproducibility±0.2%
  • AutoclavableYes
  • Wetted PartsPTFE, ETP, borosilicate glass, PP, PFA, FEP
  • DescriptionBottletop Dispenser, 10 to 100 mL
  • Warranty1 year
1 Year Warranty

Key Features

  • Recirculation valve prevents loss of reagent during priming and purging
  • Smooth plunger movement provides bubble-free dispensing
  • Adjustable nozzle allows you to change the dispensing angle
  • 1 year warranty

More About this Item

These bottletop dispensers conserve reagents with a built-in recirculation valve allowing you to prime and purge without waste. The PTFE piston with ETP o-ring provide frictionless movement reducing the occurrence of bubbles in your dispenses. The springless PTFE valve manifold offers superior chemical compatibility and resists jamming.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat