This item is now discontinued. Please browse similar items

Cole-Parmer Binocular Stereozoom Microscope; 15x-45x, Fixed-Arm, 115V

Representative image only.
Cole-Parmer Item # RK-03905-30
Stereozoom optics let you easily focus through different magnification ranges
  • Head rotates 360°> for different viewing configurations
  • Limited Lifetime Warranty

SPECS & DESCRIPTION

Specifications & Description

  • Magnification15x to 45x
  • Maximum Magnification45x
  • Head ConfigurationBinocular
  • Objectives1x to 4.5x
  • Eyepiece Magnification10x
  • EyepiecesWidefield
  • Illuminator(s)Top and bottom
  • Stand TypeFixed Arm
  • Power (VAC)115
  • DescriptionBinocular Stereozoom Microscope; 15x-45x, Fixed-Arm, 115V
  • WarrantyLTD Life Time
Limited Lifetime Warranty

Key Features

  • Head rotates 360°> for different viewing configurations
  • Limited Lifetime Warranty

More About this Item

Stereozoom microscopes feature stepless magnification through zoom control on viewing head. Head rotates 360°>; widefield 10x eyepieces feature adjustable diopter and interpupillary distance for comfortable viewing.

Fixed-arm stand provides built-in incident (top) and transmitted (bottom) illumination. Includes a 6-ft cord and three-prong plug.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat