This item is now discontinued. Please browse similar items
Cole-Parmer 96-Well Equilibrium Dialyzer - 10,000 Daltons
Representative image only.
Cole-Parmer Item # RK-02896-72
Lets you simultaneously assay up to 96 samples
  • Each well contains a separate membrane to eliminate cross-contamination
  • High reproducibility from well to well
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialRegenerated Cellulose
  • Max Molecular Weight Cut Off (Mwco) (Dalton)10000
  • Max Sample Volume (mL)0.2
  • Description96-Well Equilibrium Dialyzer - 10,000 Daltons
  • Warranty1 month
1 Month Warranty
Key Features
  • Each well contains a separate membrane to eliminate cross-contamination
  • High reproducibility from well to well
  • 1 month warranty
More About this Item
The single-use equilibrium dialyzer is a novel product for the simultaneous assay of 96 samples. Each well has a separate membrane to eliminate cross contamination. The high-quality regenerated cellulose membranes are free of sulfur and heavy metal contamination. Wells are sealed with 8-cap strips, so either a row of wells or all 96 wells can be used depending on the experiment. Reproducibility is very high across the different wells, and sample recovery is above 95%.
What’s included: twelve 8-cap top strips and twelve 8-cap bottom strips to seal wells. Rotator is required; order separately.