This item is now discontinued. Please browse similar items
Cole-Parmer 332-5727-CP Waste System, 120 mm Cap, 4 x 1/8", 1 x 1/4" OD Tube Fittings, 3 x 1/4" or 3/8" Hose Barb, Exhaust Filter; 40 L
Representative image only.
Cole-Parmer Item # RK-06064-37
Protect workers from inhaling dangerous fumes while collecting HPLC waste
  • Capture potentially dangerous solvent vapors
  • Safely collect waste with the high-density polyethylene (HDPE) carboy which features superior chemical compatibility
  • No need to remove tubing from the open cap to install a closed cap
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (L)40
  • MaterialHigh Density Polyethyene (HDPE)
  • Cap MaterialPolypropylene (PP)
  • Cap / Stopper Size120 mm
  • DescriptionCarboy Solvent Waste System, 120 mm Cap, 4 x 1/8\", 1 x 1/4\" OD Tube Fittings, 3 x 1/4\" or 3/8\" Hose Barb, Exhaust Filter; 40 L
Key Features
  • Capture potentially dangerous solvent vapors
  • Safely collect waste with the high-density polyethylene (HDPE) carboy which features superior chemical compatibility
  • No need to remove tubing from the open cap to install a closed cap
More About this Item
This heavy-duty solvent waste system is designed to collect liquid waste disposal from one or more HPLC systems while a carbon exhaust filter captures potentially harmful vapors emitted. The closed system helps eliminate spills and provides a safe and efficient way of containing, transporting, and disposing of aggressive organic solvents.
The polypropylene cap (PP) offers a wide range of tubing port configurations. The 40-L carboys come with 120 mm caps.