This item is now discontinued. Please browse similar items
Cole-Parmer 2077 Datalogger Kit with Graphing Software for Ventilation Monitor
Representative image only.
Cole-Parmer Item # RK-10101-34
Optimize performance of your air/gas analyzer
  • Kit comes with everything you need to record and graph CO2, relative humidity and temperature
  • Typically used with monitor but can also function as a stand-alone product
  • Compact 4-channel data logger has RH and temperature sensors built in and two external channels that accept V or mA signals
  • Nonvolatile EEPROM memory retains data even if battery fails
  • Deluxe software (included) for launching and retrieving the data logger and graphing the data
  • Data can easily be exported to other spread sheet programs like Microsoft® Excel®
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionRh/Temp Datalogger Kit w/ Graphing Software Custom Interface Cables
  • Warranty1 year
1 Year Warranty
Includes
data logger with built-in RH and temperature sensors, PC graphing software, and cables.
Key Features
  • Kit comes with everything you need to record and graph CO2, relative humidity and temperature
  • Typically used with monitor but can also function as a stand-alone product
  • Compact 4-channel data logger has RH and temperature sensors built in and two external channels that accept V or mA signals
  • Nonvolatile EEPROM memory retains data even if battery fails
  • Deluxe software (included) for launching and retrieving the data logger and graphing the data
  • Data can easily be exported to other spread sheet programs like Microsoft® Excel®
  • 1 year warranty
More About this Item
Replacement sensors, carrying cases, and other accessories to customize your sampling and analysis system.
Compatible with air analysis monitors 86290-00 and -10.