This item is now discontinued. Please browse similar items
Cole-Parmer 1340-5354 Handheld Digital Vacuum Gauge, 0 to 1999 mbar
Representative image only.
Cole-Parmer Item # RK-68980-02
Use optional extension cables to take remote pressure measurements
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±0.4% full-scale
  • Wetted MaterialsPEEK
  • CaseABS plastic
  • Process Connection1/4\" to 3/8\" OD tubing
  • Unit Of Measurementmbar
  • Display3.5-digit LCD
  • Battery TypeRechargeable NiCd battery (included)
  • Media CompatibilityGases and liquids compatible with wetted materials
  • Resolution1 mbar
  • DescriptionHandheld Digital Vacuum Gauge, 0 to 1999 mbar
  • Warranty2 year
Two year warranty
Key Features
  • 2 year warranty
More About this Item
Our compact, lightweight gauge is ideal for vacuum system use--measures vacuum from 0 to 2000 mbar. Durable ceramic sensor is enclosed in polyethyletherketone (PEEK) housing to offer excellent chemical compatibility and provide years of reliable service. Sensor is detachable--order optional extension cables to take remote readings up to 16 feet (5 meters) from gauge.
The ABS plastic housing resists many chemicals. Rear-mounted clip makes it easy to carry or mount the gauge. Order optional adapters to use gauge with flanged or glass connections.