This item is now discontinued. Please browse similar items
Cole-Parmer 0.01 To 20 Torr Pressure/vacuum Pirani-Type Sensor, NW 16 Connection
Representative image only.
Cole-Parmer Item # RK-68801-42
Customize to your needs
  • Complete system requires:
  • 1. Sensor
  • 2. Indicator or controller
  • Optional 3. Calibrator
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±5% of reading or ±0.03 Torr
  • Wetted Materials304 SS
  • Process ConnectionNW 16 flange
  • Media CompatibilityGases compatible with wetted materials
  • Resolution0.01 Torr
  • Description0.01 To 20 Torr Pressure/vacuum Pirani-type Sensor, Nw 16 Connection
  • Warranty1 year
1 Year Warranty
Key Features
  • Complete system requires:
  • 1. Sensor
  • 2. Indicator or controller
  • Optional 3. Calibrator
  • 1 year warranty
More About this Item
Sensors provide improved tracking ability and maintain calibration over long periods of time. When replacing a sensor, you do not need to recalibrate.
These sensors are suited for use with mercury and oil pumps.
Pirani type sensors offer enhanced stability and an automatic temperature compensation network. The 304 stainless steel sensors operate at half the
temperature of standard thermopile sensors. Use for high-vacuum applications.
Diaphragm type sensor offers quick response—less than 400 milliseconds full-scale. Select a diaphragm type sensor made from 316 stainless steel or silicon based on your application.