This item is now discontinued. Please browse similar items
CELLTREAT Scientific Products 229682 Dual Compartment Sterile Petri Dishes, 100 x 15 mm; 500/cs
Representative image only.
CELLTREAT Scientific Products Mfr # 229682 – Item # RK-01932-22
Optically clear for improved imaging and observation
  • Consistent flatness allows for even media distribution
  • Manufactured from virgin polystyrene
  • All dishes are supplied sterile (gamma irradiated)
  • Run multiple experiments in one petri dish
  • 7 to 8 mL working volume/compartment
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDual Compartment Sterile Petri Dishes, 100 x 15 mm; 500/cs
Sterile
Key Features
  • Consistent flatness allows for even media distribution
  • Manufactured from virgin polystyrene
  • All dishes are supplied sterile (gamma irradiated)
  • Run multiple experiments in one petri dish
  • 7 to 8 mL working volume/compartment
More About this Item
Use the compartment dishes to run simultaneous studies of organisms in a common environment. Lids have molded spacers to reduce condensation and allow air circulation. Stacking rings on the lids provide increased stability.
All dishes are supplied sterile (gamma irradiated) and come packaged 25 dishes/bag.