CELLTREAT Scientific Products 229461 Oak Ridge Centrifuge Tube, 30 mL, Screw Cap, Bag, Nonsterile; 12/Cs
Representative image only.
CELLTREAT Scientific Products Mfr # 229461 – Item # RK-06336-26
Ideal for high-speed, refrigerated, or standard centrifugation
  • Featuring leak-proof screw caps, uniform wall thickness, all-polypropylene construction, and round bottoms—these tubes can withstand up to 50,000 RCF x g
PRODUCT OPTIONS
$108.29CAD / Case of 12
SPECS & DESCRIPTION
Specifications & Description
  • Max RCF x g50000
  • SterileNo
  • Tube Capacity (mL)30
  • Tube MaterialPolypropylene
  • Tube ColorOpaque
  • Tube Bottom ShapeRound/Autoclavable
  • Cap IncludedYes
  • Cap StyleScrew Cap
  • GraduationsNo
Key Features
  • Featuring leak-proof screw caps, uniform wall thickness, all-polypropylene construction, and round bottoms—these tubes can withstand up to 50,000 RCF x g