This item is now discontinued. Please browse similar items
Casella P109032 Low Flow Module for Tuff Personal Sampling Pump
Representative image only.
Casella Mfr # P109032 – Item # RK-86543-43
Tough, rugged, and reliable for sampling in difficult environments
  • High-intensity run/fault LEDs for error condition warning
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionLow Flow Module for Tuff Personal Sampling Pump
  • Warranty1 year
1 Year Warranty
Key Features
  • High-intensity run/fault LEDs for error condition warning
  • 1 year warranty
More About this Item
Rubber over-molding, reinforced LCD window, and full gasket seal make this the product of choice in tough environments. Foam filter in the inlet port minimizes entry of fine dust particles. Simple four-button operation allows quick and easy setup. Set flow rates down to 5 mL/min with the optional low-flow adapter — great for use with sorbent tubes.