This item is now discontinued. We recommend the following as suggested replacements: RK-06101-40
Burkle 0319-0250 Sample Bottle, Polypropylene, Transparent; 250 mL
Representative image only.
Burkle Mfr # 0319-0250 – Item # RK-06040-16
Accessories for the vacuum-operated liquid sample collection
  • Reach narrow, less-accessible areas with the small-diameter hose
  • Avoid contamination with the easily replaceable suction hose and sample bottle
  • 15 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Wetted MaterialsPolypropylene
  • DescriptionSample bottle, PP, 40 mm cap, transparent; 250 mL
  • Warranty15 month
15 Month Warranty
Key Features
  • Reach narrow, less-accessible areas with the small-diameter hose
  • Avoid contamination with the easily replaceable suction hose and sample bottle
  • 15 month warranty
More About this Item
These accessories are suitable for use with the Bürkle industrial vacuum pumps for sealed liquid sampling.
The aluminum sampler (model 06040-99) is ideal for sampling solvents and flammable liquids in explosion categories IIA, IIB, and IIC. This model comes with an earthing cable and electrically conductive tubing for sampling of these liquids. This sampler can be used with sampling bottles 06041-29 or 06041-30 and suction tube 06040-98.
The polypropylene sampler (model 06040-97) is ideal for general liquid sampling of weak acids, alkalis, detergents, and chemical baths. This sampler can be used sampling bottle 06040-16 and suction hose 06040-98.