This item is now discontinued. Please browse similar items
Bud Industries 30-2301-GT Instrument Cabinet, 40" high.
Representative image only.
Bud Industries Mfr # 30-2301-GT – Item # RK-94501-52
Enclosures provide high-quality storage for instruments, controllers, and electronics
  • Ships completely assembled
  • Mount any 19” wide instrument directly to enclosure
  • Shelves provide heavy-duty surface for smaller instruments or electronics
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialSteel
  • Outside Height (in)40
  • DescriptionInstrument Cabinet, 40\" high.
  • Warranty3 Months
3 Month Warranty
Key Features
  • Ships completely assembled
  • Mount any 19” wide instrument directly to enclosure
  • Shelves provide heavy-duty surface for smaller instruments or electronics
  • 3 month warranty
More About this Item
Model 94501-52 has a frame constructed of 14-gauge steel with U shaped 16-gauge steel panel-mounting rail supports. Cabinet has a 1200 lb load capacity. Side and top panels are constructed of 18-gauge steel. One door is furnish and can be used in the front or back of the cabinet. Door is constructed of 20-gauge steel.