This item is now discontinued. Please browse similar items
BRAND PLT™ Pipette Leak Testing Unit (PLT Unit), Single Channel; 100-240 VAC/50-60 Hz
Representative image only.
Mfr # 703970 – Item # RK-24550-05
A quick, daily functional tester for pipettes between calibrations
  • Test pipettes with or without tips
  • Detects even the smallest of leaks
  • Speedy results with pass or fail criteria in seconds
  • 1 year warranty
Often Bought With…
SPECS & DESCRIPTION
Specifications & Description
  • Min Volume (µL)1
  • Max Volume (mL)10
  • Number Of Channels1
  • Auto ShutoffCustomizable 1 to 30 minute
  • Filter TypePolyethylene
  • Power (VAC)100 to 240
  • Compatible withPiston-Operated Pipettes
  • DescriptionPipette Leak Testing Unit, Single Channel, 1 µL to 10 mL; 100-240 VAC, 50/60 Hz
  • Warranty1 year
1 Year Warranty
Key Features
  • Test pipettes with or without tips
  • Detects even the smallest of leaks
  • Speedy results with pass or fail criteria in seconds
  • 1 year warranty
More About this Item
Ensure quality results with the BRAND® PLT™ pipete leak tester. Over 90% of pipette failures are due to leakages. The PLT™ unit quantifies leakages, and determines whether that leakage will make a gravimetricaly detectable error. The PLT™ unit can also be used as a diagnostic tool, helping to narrow down the source of the leakage, and can even help verify the fit of 3rd party pipette tips.