This item is now discontinued. Please browse similar items

BrandTech 115015 Sample Cup for Technicon® Analyzers, PS, 1.5 mL; 12,000/PK

Representative image only.
BrandTech Mfr # 115015 – Item # RK-06101-34
Designed to fit common analyzers
  • Clear polystyrene sample cups provide quick and acurate analysis
  • For Technicon® analyzers
  • 1.5 mL capacity
  • 15 mm top dia x 12.2 mm bottom dia

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)1.5
  • MaterialPolystyrene
  • ColorClear
  • Compatible WithTechnicon® analyzers
  • Height (mm)22.7
  • DescriptionSample Cup for Technicon® Analyzers, PS, 1.5 mL; 10,000/PK

Key Features

  • Clear polystyrene sample cups provide quick and acurate analysis
  • For Technicon® analyzers
  • 1.5 mL capacity
  • 15 mm top dia x 12.2 mm bottom dia

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat