This item is now discontinued. Please browse similar items
Brady X-120-499 Label Cartridge, 1"W, Nylon
Representative image only.
Brady Mfr # X-120-499 – Item # RK-65544-22
Pre-sized labels customized to fit vials, tubes, and slides
  • Built-in bar coding
  • One-touch time and date stamping
SPECS & DESCRIPTION
Specifications & Description
  • Accessory TypeLabel Cartridge
  • Label size25.4 mm W x 12.7 mm H
  • DescriptionLabel Cartridge, 1\"W, Nylon
Key Features
  • Built-in bar coding
  • One-touch time and date stamping
More About this Item
Quickly and easily label your vials, vial tops, slides, well plates, and tubes with the Labxpert 300-dpi thermal transfer label printer. Printer features a QWERTY keyboard, easy-to-use function buttons, and a large, easy-to-read LCD. Unit prints over 140 lab-specific Greek, research, and safety symbols, as well as built-in bar coding.
Labels are designed for harsh laboratory environment — they won't fall off, crack, or peel, and labels are designed not to get jammed in centrifuges or other equipment. Labels can withstand extreme temperatures, such as liquid nitrogen (-196°C) through autoclave (+121°C). Labels also withstand harsh chemicals, such as xylene, ethanol, DMSO, IPA, and toluene. To choose the correct label for your application, refer to the special properties and label diagram columns. Operates with AC power supply or AA batteries.