This item is now discontinued. Please browse similar items
Brady BMP-71 BMP 71 Laboratory Label Printer with Battery Pack and AC Adapter
Representative image only.
Brady Mfr # BMP-71 – Item # RK-65533-32
Large, graphical, full color display with on screen indicators
  • Stand alone or PC compatible
  • Date/Time stamp
  • Bar code symbologies
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionBMP 71 Laboratory Label Printer with Battery Pack and AC Adapter
Conformite Europeenne
Key Features
  • Stand alone or PC compatible
  • Date/Time stamp
  • Bar code symbologies
More About this Item
This portable thermal transfer printer features 300 dpi printing, prints 1.5" per second, and features a QWERTY style keypad layout with built-in touch cell allowing printer to automatically format label. Printer can stand alone, is PC compatible, and features a large, graphical, full-color display. Dispay indicates installed label part, label size and color, label and area status, current font size and rotation, amount of material remaining, and remaining battery charge.
Additionally, printer can store, retrieve and delete label files — save more than 100,000 label files. Print a batch of labels or multiple copies of one label. Printer uses Brady LabelMark label design software and has a sleep mode when not in use to conserve energy.