This item is now discontinued. Please browse similar items

Brady 105961 Personal Electrical Lockout Kit in Plastic Case

Representative image only.
Brady Mfr # 105961 – Item # RK-86700-01
Provides lock out of circuit breakers, electrical plugs and disconnect switches
  • Contents
  • Mini Cable lockout
  • 3-in-1 plug lockout
  • Oversized breaker lockout
  • Two 120/277 V clamp-on breaker lockouts with cleats
  • One 480/600 V clamp-on breaker lockout with cleat
  • Two universal multi-pole breaker lockouts
  • Three 1" (2.5 cm) group lockou

SPECS & DESCRIPTION

Specifications & Description

  • DescriptionPersonal Electrical Lockout Kit in Plastic Case

Key Features

  • Contents
  • Mini Cable lockout
  • 3-in-1 plug lockout
  • Oversized breaker lockout
  • Two 120/277 V clamp-on breaker lockouts with cleats
  • One 480/600 V clamp-on breaker lockout with cleat
  • Two universal multi-pole breaker lockouts
  • Three 1" (2.5 cm) group lockou

More About this Item

This lockout kit provides a variety of devices to lock out circuit breakers, electrical plugs, and disconnect switches. Kit comes in a durable plastic case, and contains everything needed to comply with OSHA.

What's included:

* Mini Cable lockout

* 3-in-1 plug lockout

* Oversized breaker lockout

* Two 120/277 V clamp-on breaker lockouts with cleats

* One 480/600 V clamp-on breaker lockout with cleat

* Two universal multi-pole breaker lockouts

* Three 1" (2.5 cm) group lockout hasps

* Three heavy-duty lockout tags with nylon cable ties

* Three keyed-alike safety products

Compliance: OSHA 29 CFR Part 1910.147

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat