This item is now discontinued. Please browse similar items
Botulinum Toxin Detection Kit, handheld assay, 10/bx
Representative image only.
Item # RK-36108-14
Quickly determine biological threat credibility
  • Results in 3- 30 minutes
  • Easy to read – two lines for positive; one line for negative
  • High Sensitivity and specificity
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Test TypeBotulinum Toxin
  • Number of Tests Included10
  • DescriptionBotulinum Toxin Detection Kit, handheld assay, 10/bx
  • Warranty2 year
Two year warranty
Key Features
  • Results in 3- 30 minutes
  • Easy to read – two lines for positive; one line for negative
  • High Sensitivity and specificity
  • 2 year warranty
More About this Item
Accurately detect small, yet high threat amounts of biowarfare agents-without extensive sample preparation or additional expensive equipment. Test powder or liquid samples; kits are simple to use, results are quick and easy to read. Biohazard disposal bags and chain of custody labels are included for proper control and disposal or transport of the sample and test. Order simulation devices below for training purposes.
What’s included: 5" x 7" pouch includes all material to prepare and test a sample as well as package it for disposal or transport after testing.