This item is now discontinued. Please browse similar items
Bio-Works BabyBio Q Pre-Packed Chromatography Column, Quaternary Amine; 10 x 5 mL
Representative image only.
Bio-Works Mfr # KN45100108 – Item # RK-11947-53
Ready to use for quick and easy purification of proteins
  • Rapid method screening and separation
  • Agarose-based resin provides optimal selectivity, resolution, and purity
  • Applications: Strong anion exchange, 3 to 12 pH
SPECS & DESCRIPTION
Specifications & Description
  • Column Volume (mL/cm)5
  • Wetted MaterialsQuaternary amine resin
  • Total Volume Capacity (mL)Dynamic binding: 50 mg BSA/mL medium
  • ApplicationsStrong anion exchange, 3 to 12 pH
  • DescriptionPre-Packed Chromatography Column, Quaternary Amine; 10 x 5 mL
Key Features
  • Rapid method screening and separation
  • Agarose-based resin provides optimal selectivity, resolution, and purity
  • Applications: Strong anion exchange, 3 to 12 pH
More About this Item
BabyBio IEC columns come prepacked with WorkBeads™ media, agarose-based chromatographic resins, which provide a tight size distribution and exceptional mechanical stability. Beads in columns maintain high capacity over broad pH ranges. Connect up to five columns in series to process larger sample volumes.
Columns feature biocompatible polypropylene body and polyethylene top and bottom filters. Each column comes with a plug in the inlet and a snap-off end at the outlet. Easily connect to a syringe, pump, or chromatography system using fingertight fittings (coned 10-32) for 1/16" OD tubing.