This item is now discontinued. Please browse similar items
Martel 1919919 Handheld Pressure Calibrator Kit; Electric/300 psi
Representative image only.
Martel Mfr # 1919919 – Item # RK-68510-16
Electric Pump Generates Pressure to 150 psi
  • The 330 comes in 3 ranges of 30 PSI, 150 PSI and 300 PSI. The 300 PSI version uses a manually operated handpump.
  • The compact, rugged design operates on eight (8) standard AA Alkaline batteries.
  • Performs approximately 300 calibration cycles to 150 PSI on one set of batteries.
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionHandheld Pressure Calibrator Kit; Electric/300 psi
  • Warranty1 Year
1 Year WarrantyConformite Europeenne
Key Features
  • The 330 comes in 3 ranges of 30 PSI, 150 PSI and 300 PSI. The 300 PSI version uses a manually operated handpump.
  • The compact, rugged design operates on eight (8) standard AA Alkaline batteries.
  • Performs approximately 300 calibration cycles to 150 PSI on one set of batteries.
  • 1 year warranty
More About this Item
The new BetaGauge 330 Pressure Calibrator provides a number of convenience features. Switch test can be performed on internal or external pressure input. Percent (%) Error and damping functions are provided. Up to five frequently used setups can be stored and retrieved with one button recall.
A Pt100 RTD input is provided for temperature measurements accurate to ±0.1 °C (0.2 °F).
In addition, the BetaGauge 330 measures and simulates 4-20 mA loop current signals. It can measure up to 30 VDC. An internal 24 VDC Instrument Power Supply supplies power to a transmitter under test.