Bel-Art Robotic Sterile Filtered Pipette Tips, Hamilton Type, 1000uL, Non-Conductive; 96 Tips/Rack x 16/PK
Representative image only.
Bel-Art Mfr # 378601100 – Item # RK-07898-06
Ideal for use with Hamilton® and Tecan® robotic systems
  • DNase, RNase, and pyrogen free
  • Manufactured in an ISO Class 8 cleanroom
  • Filtered to prevent contamination of pipette arm
  • 3 month warranty
PRODUCT OPTIONS
$586.41CAD / Pkg of 16
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Max Volume (µL)50
  • Max Volume (mL)0.05
  • Tip StyleNon-conductive
  • Body MaterialClear Polypropylene
  • Color CodeNatural
  • Package Of16 racks; 96 tips/rack
  • Compatible withHamilton robotic systems
  • DescriptionRobotic Sterile Filtered Pipette Tips, Hamilton Type, 1000uL, Non-Conductive; 96 Tips/Rack x 16/PK
  • Warranty3 month
3 Month WarrantyConformite Europeenne
Key Features
  • DNase, RNase, and pyrogen free
  • Manufactured in an ISO Class 8 cleanroom
  • Filtered to prevent contamination of pipette arm
  • 3 month warranty
More About this Item
These sterile, disposable, filtered pipette tips can be use with automated liquid handling equipment as well as with Hamilton® and Tecan® robotic systems. The filtered tips protect pipette arm from cross-contamination.
Available in conductive and non-conductive tips. The conductive tips detect the liquid level when pipetting conductive liquids to ensure repeatable volume accuracy.