This item is now discontinued. Please browse similar items
Bel-Art F65000-9005 Tube and Pestle, 0.5 mL, Bulk Packed; 100/Pk
Representative image only.
Bel-Art Mfr # F65000-9005 – Item # RK-44444-88
Process precipitates, pellets, tissue, and cells directly in the microtube!
  • Homogenize samples directly in their storage container
  • Obtain thorough mixtures with the tight pestle-to-microtube tolerance
  • Protect your samples—tubes and pestles are made from virgin polypropylene
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)0.5
  • MaterialPolypropylene
  • Description Tube and Pestle, 0.5 mL, Bulk Packed; 100/Pk
  • Warranty3 month
3 Month Warranty
Key Features
  • Homogenize samples directly in their storage container
  • Obtain thorough mixtures with the tight pestle-to-microtube tolerance
  • Protect your samples—tubes and pestles are made from virgin polypropylene