Bausch & Lomb Coddington Magnifier, 10X, 0.78" Diameter
Representative image only.
Bausch & Lomb Mfr # 81-61-31 – Item # RK-03883-10
Provide good definition and a wide field of view
  • Durable nickel-plated case protects the lens
  • 1 year warranty
$76.47CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Magnification10x
  • Viewing Area0.78\" (19.8 mm) diameter
  • LensSingle
  • Focal Length1\" (25.4 mm)
  • BatteryNone
  • DescriptionCoddington Magnifier, 10x Magnification
  • Warranty1 Year
1 Year Warranty
Key Features
  • Durable nickel-plated case protects the lens
  • 1 year warranty
More About this Item
These high-performance Coddington magnifiers provide sharp, crisp images. Correction is achieved through the use of a single thick lens with a central groove diaphragm. Swing-away, nickel-plated case protects the lens and acts as a handle.