This item is now discontinued. Please browse similar items
Bausch & Lomb Folding Pocket Magnifier, Triple Lens, 5X to 20X
Representative image only.
Bausch & Lomb Mfr # 812367 – Item # RK-03882-19
Hard plastic case protects the lens in the field and in the lab
  • Durable, handheld magnifer is portal for a wide range of applications
  • Triple lens features 5X to 20X magnification
SPECS & DESCRIPTION
Specifications & Description
  • Magnification5x to 20x
  • LensTriple
  • Focal Length2\" to 0.5\" (50.8 mm to 12.7 mm)
  • DescriptionFolding Pocket Magnifier, triple lens, 5x to 20x magnification
Key Features
  • Durable, handheld magnifer is portal for a wide range of applications
  • Triple lens features 5X to 20X magnification
More About this Item
Durable plastic swing-away case protects the lens and acts as a handle during use. For models with more than one lens, combine lens for different magnification power.