This item is now discontinued. Please browse similar items
Bausch & Lomb Hastings Triplet Magnifier, 7X, 0.78" Diameter
Representative image only.
Bausch & Lomb Mfr # 81-61-68 – Item # RK-03882-20
Provides sharp, distinct images without distortion
  • Lenses are recessed to protect it from damage
  • Swing-away, nickel-plated case also protects the lens and acts as a handle
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Magnification7X
  • Viewing Area0.78\" (19.8 mm) diameter
  • LensTriple
  • Focal Length1.5\" (38.1 mm)
  • DescriptionHastings Triplet Magnifier, 7X, 0.78\" Diameter
  • Warranty1 Year
1 Year Warranty
Includes
protective storage pouch.
Key Features
  • Lenses are recessed to protect it from damage
  • Swing-away, nickel-plated case also protects the lens and acts as a handle
  • 1 year warranty
More About this Item
These magnifiers eliminate the distortion commonly found with similarly priced pocket magnifiers by bonding together three optical lenses, forming a "Hastings Triplet". Hastings Triplet Magnifiers incorporate three separate glass lenses bonded together to form a compound lens to provide sharp, very distinct magnified images without distortion.