This item is now discontinued. Please browse similar items
Ashcroft LTA Temperature Switch Remote Mount, 5' Capillary, 20/95°C, Adjustable Deadband w/ Adapter
Representative image only.
Ashcroft Mfr # LTA N4 H 05-A7-030 XBX 20/95C – Item # RK-39011-37
Prevent unneeded activation with adjustable deadband
  • Quick, accurate response via SAMA Class II vapor pressure thermal system
  • Choose the switch to meet your needs—wide variety of lengths and temperature ranges available
  • Easy to operate and install—meet NEMA 4, 4X, and IP66 requirements
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Process Connection1/2\" NPTM Adapter
  • Electrical Connections(1) 3/4\" NPTF
  • Min Temperature (° C)20
  • Temperature Accuracy±1%
  • Height (in)11 9/16
  • Width (in)4 1/8
  • DescriptionTemperature Switch Remote Mount, 5' Capillary, 20/95°C, Adjustable Deadband w/ Adapter
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Quick, accurate response via SAMA Class II vapor pressure thermal system
  • Choose the switch to meet your needs—wide variety of lengths and temperature ranges available
  • Easy to operate and install—meet NEMA 4, 4X, and IP66 requirements
  • 1 year warranty
More About this Item
Single set point, adjustable deadband, SPDT switches feature epoxy-coated aluminum systems for reliable performance. Choose direct-mounting units with rigid probes—great for use with 3/8" nominal bore thermowells in areas with high vibration, velocity, or pressure. Remote-mount units have spring-mounted capillaries with 302 stainless steel armor and include a 1/2" NPT(M) bushing adapter.