This item is now discontinued. Please browse similar items
Argos Technologies Pipetting Work Station Inserts, 12x1.5mL, Purple; 5/Pk
Representative image only.
Argos Technologies Item # RK-22999-44
  • Interchangeable, inserts can also be used as stand-alone racks
SPECS & DESCRIPTION
Specifications & Description
  • Max Tube Size (mL)1.5
  • Tubes Held12
  • MaterialPolypropylene(PP)
  • ColorPurple
  • DescriptionPipetting Work Station Inserts, 12x1.5mL, Purple; 5/Pk
Key Features
  • Interchangeable, inserts can also be used as stand-alone racks
More About this Item
Color-coded polypropylene inserts accept tubes with sizes ranging from 0.5 to 50 mL. Easy-release tabs allow you to quickly change out the inserts to customize the rack to your applications. Each tube cavity is numbered for quick identification. Use the pipetting basin insert to hold up to 25 mL of reagent or solution.