This item is now discontinued. Please browse similar items
Argos Technologies Pipette Tip Box, Polycarbonate, 1000ul, 128-Place Grid Insert; Blue Rack
Representative image only.
Argos Technologies Item # RK-24406-66
Ideal for pipette tip protection and storage
  • Transparent box contains a hinged lid for easy one hand opening
  • Autoclavable at 121°C
  • Constructed of durable polycarbonate
  • Accomidates 1000 μl pipette tips
  • Features a 128-place grid insert for quick organization
  • Dimensions of 6-3/4" L x 3-3/16" H x 3-9/16" W
  • Designed with transparent box and blue pipette rack
SPECS & DESCRIPTION
Specifications & Description
  • Width (InchShort)3 9/16
  • Height (in)3 3/16
  • AutoclavableYes
  • Overall Length (in)6.75
  • Wetted MatterialPolycarbonate
  • Color CodeBlue
  • Compatible with129 x 1000 ul Pipette Tipe
  • DescriptionPipette Tip Box, Polycarbonate, 1000ul, 128-Place Grid Insert; Blue Rack
Key Features
  • Transparent box contains a hinged lid for easy one hand opening
  • Autoclavable at 121°C
  • Constructed of durable polycarbonate
  • Accomidates 1000 μl pipette tips
  • Features a 128-place grid insert for quick organization
  • Dimensions of 6-3/4" L x 3-3/16" H x 3-9/16" W
  • Designed with transparent box and blue pipette rack
More About this Item
Choose from pipette storage boxes with included pipette tip rack or storage box with removable divider.