This item is now discontinued. Please browse similar items
Argos Technologies Pellet Mixer Microtube, 0.5 mL; 100/Pk
Representative image only.
Argos Technologies Item # RK-44468-22
Quickly homogenize cells, plant tissue, and DNA pellets within microtube vials
  • Free of DNAse, RNAse, and pyrogens to eliminate sample contamination
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)0.5
  • Capacity (Liters)0.0005
  • DescriptionPellet Mixer Microtube, 0.5 mL; 100/Pk
Key Features
  • Free of DNAse, RNAse, and pyrogens to eliminate sample contamination
More About this Item
Plastic pestles and tubes are designed to homogenize cells and plant tissue or to resuspend protein and DNA pellets. They are DNAase/RNAase and pyrogen free, and come individually wrapped to guarantee purity. The pestle features a grooved grip, making it easy to turn without slipping, even while wearing gloves. Pestle length is 3” (7.5 cm). Both pestle and tube are autoclavable. Compatible with model 44468-25 pestle mixer, order separately.