This item is now discontinued. Please browse similar items

Argos Technologies Nova® Microcinerator Replacement Fuse, 1.25 Amp; 220 VAC

Representative image only.
Argos Technologies Item # RK-24410-47
Enhance the versitility of your Nova® Microcinerator Loop and Needle Sterilizer

SPECS & DESCRIPTION

Specifications & Description

  • Compatible WithArgos Nova Microincinerator; 220 VAC
  • Power (VAC)220
  • Power (amps)1.25
  • DescriptionMicrocinerator Replacement Fuse, 1.25 Amp; 220 VAC

More About this Item

The slide dryer is a safe and easy way to use heat already generated by the incinerator to dry slides. This is ideal for dissolving epithelium using KOH mycology technique.
The loop rest protects loop handles from overheating or melting. This accessories fits any loop and attaches easily to the incinerators.
The heating element of your Nova® Microcinerator Loop and Needle Sterilizer will last approximatley 6 months depending on usage. Easily and quikly replace these heating elements to keep your incinerator up and running. Model 04397-12 is for the 120V units and 04397-16 is for the 220V powered incinerators.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat