This item is now discontinued. Please browse similar items
Argos Technologies Manual Pipette Pump, 2 mL, Blue
Representative image only.
Argos Technologies Item # RK-04395-04
Economical choice for accurate dispensing
  • Resists acids and alkalis
  • Color coded for easy identification
  • Unit disassembles for easy cleaning
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Max Volume (mL)2
  • Color CodeBlue
  • DescriptionManual Pipette Pump, 2 mL, Blue
Key Features
  • Resists acids and alkalis
  • Color coded for easy identification
  • Unit disassembles for easy cleaning
More About this Item
Manual pipette pumps feature a thumbwheel for precision filling or dispensing of liquids. Insert glass or plastic pipettes into secure tapered collar and quickly dispense liquid by depressing white side bar or plunger. Operate easily with one hand.