Cole-Parmer Essentials Disposable Serological Pipettes, 10 mL, Individually Wrap; 200/Cs
Representative image only.
Cole-Parmer Essentials Item # RK-04395-21
Filter plug prevents overfilling
  • Color coded rings ease identification
  • Constructed from transparent medical-grade polystyrene
  • Sterilized by gamma irradiation and tested before lots are released
PRODUCT OPTIONS
$128.18CAD / Case of 200
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Max Volume (mL)10
  • Graduation Interval0.1 mL
  • Body MaterialPolystyrene
  • Color CodeOrange
  • DescriptionDisposable Serological Pipettes, 10 mL, Individually Wrap; 200/Cs
Key Features
  • Color coded rings ease identification
  • Constructed from transparent medical-grade polystyrene
  • Sterilized by gamma irradiation and tested before lots are released
More About this Item
Designed for accurate and rapid fluid dispensing, these disposable polystyrene (PS) serological pipettes feature a filter plug which eliminates lint fibers and a uniform mouth piece. Sharp, black, permanent graduations guarantee precise reading and accurate dispensing to ±2%. Reverse graduations show volume removed or remaining on all pipettes larger than 1 mL. Negative graduations provide additional pipetting volume.
Pipette with 1.1-mL capacity meets the APHA standards for the examination of dairy products.
The pipettes come individually wrapped in a fiber-free paper with easy peel-apart opening. Color-coded dispenser box allows quick access to pipettes without opening top.