This item is now discontinued. Please browse similar items
Applikon RAMbio Z861050010 Clamp for 500 mL Flask, with screws
Representative image only.
Applikon RAMbio Mfr # Z861050010 – Item # RK-29206-85
Optimize performance of your Applikon RAMbio® Benchtop Acoustic Mixer
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Platform Flask Capacity1 x 500 mL flask
  • DescriptionClampfor500mLFlask,withscrews
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Mixer accommodates flasks ranging from 250 mL to 1 L (order mixer 29206-83 separately).
Use the RAMbio mixer in conjunction with the Oxy-Pump stopper to achieve bioreactor-equivalent oxygen transfer. The stopper actively pumps air into and out of your flask cultures. It offers complete headspace refreshment equivalent to that achieved in stirred tank bioreactors without the need for mechanical pumps. This autoclavable stopper is constructed from silicone and polypropylene and has a replaceable filter.