This item is now discontinued. Please browse similar items
Applikon Optional Glass Bottle for Fermentation Sampling System; 30 mL
Representative image only.
Applikon Mfr # Z81207BT30 – Item # RK-01919-43
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)30
  • MaterialGlass
  • Compatible With01919-19 Sampling System
  • DescriptionOptional Glass Bottle for Fermentation Sampling System; 30 mL
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Sampling system easily lets you remove a 60-mL sample from your fermentation vessel without removing the headplate. Attach system to the sample pipe via 1/4" ID tubing. Includes a 60-mL glass bottle with stainless steel fittings; optional 30-mL bottle is available, as well as additional 60-mL bottles.