Applikon Heat Exchanger for Fermentation/Bioreactor System with 10-mm Size Port, 15 L
Representative image only.
Applikon Mfr # Z81317HE10 – Item # RK-01919-83
  • 1 year warranty
$6,161.72CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithCole-Parmer Modular Fermentation Systems
  • DescriptionHeat Exchanger for Fermentation/Bioreactor System with 10-mm Size Port, 15 L
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Heat exchangers circulate warm or cold water to maintain vessel tempperature. Not recommended for cell culture systems.