This item is now discontinued. Please browse similar items

Applikon Fermentation Sampling System with 60 mL Bottle

Representative image only.
Applikon Mfr # Z81207SS02 – Item # RK-01919-19
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)60 or 30 mL; comes with 60 mL bottle
  • MaterialGlass and Stainless Steel
  • DescriptionFermentation Sampling System with 60 mL Bottle
  • Warranty1 year
1 Year Warranty

Key Features

  • 1 year warranty

More About this Item

Sampling system easily lets you remove a 60-mL sample from your fermentation vessel without removing the headplate. Attach system to the sample pipe via 1/4" ID tubing. Includes a 60-mL glass bottle with stainless steel fittings; optional 30-mL bottle is available, as well as additional 60-mL bottles.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat