This item is now discontinued. Please browse similar items
Applikon Assembly Holder with O-Ring for 10-mm Size Port
Representative image only.
Applikon Mfr # Z81320AH00 – Item # RK-01919-17
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionAssembly Holder with O-Ring for 10-mm Size Port
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Assembly holder easily accommodates 6-mm piping into your fermentation system. Simply screw this assembly holder into a 10-mm port and connect the 6-mm tubing. Includes silicone O-ring.