Ansell TouchNTuff® 92-575 Powdered Disposable Nitrile Gloves, Size S (6 ,5-7); 100/Box
Representative image only.
Ansell Mfr # 105125 BOX – Item # RK-81610-40
Keep hands dry and protected from chemical splashes
  • Textured fingertips provide secure grip
  • Complies with FDA food handling requirements
  • Ambidextrous
PRODUCT OPTIONS
$35.07CAD / Box of 100
SPECS & DESCRIPTION
Specifications & Description
  • Thickness (mil)5.5
  • MaterialNitrile
  • SizeSmall
  • Length (in)9 1/2
  • Length (cm)24
  • ColorBlue
  • GripTextured fingertips
  • CuffStandard
  • ComplianceFDA 21 CFR 177-2600
  • SterileNo
  • PowderedYes
  • DescriptionPowdered Disposable Nitrile Gloves, Size S (6,5-7)
Key Features
  • Textured fingertips provide secure grip
  • Complies with FDA food handling requirements
  • Ambidextrous
More About this Item
These robust nitrile gloves provide added protection for food processing and heavy-duty jobs. Powdered for easier donning and moisture absorption, the gloves are ideal for transferring liquids and solids, processing samples, handling incoming good, maintenance, and applications that have food contact.