This item is now discontinued. Please browse similar items
Ansell 950193 56-910 Vinyl Coat Apron; Thickness, 8-mil; Size, X-large
Representative image only.
Ansell Mfr # 950193 56-910 – Item # RK-81620-58
Also protects shoulders and arms
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)57
  • Length (cm)144.78
  • MaterialVinyl
  • SizeX-Large
  • ColorBlue
  • Thickness (mil)8
  • DescriptionVinyl Coat Apron; Thickness, 8-mil; Size, X-large
More About this Item
These coat aprons are made of FDA compliant materials — ideal for use in food handling applications. Seamless design eliminates contaminant traps while permitting cleaning to Class 100 standards. Coat aprons feature an open back/sleeves, no elastic, and have 40-mil PVC grommets permanently welded to the apron for greater strength and durability.