This item is now discontinued. Please browse similar items
Ansell 565718 Blue Nitrile Gloves, Powder Free, Large
Representative image only.
Ansell Mfr # 565718 – Item # RK-81601-06
Well suited for light-duty maintenance, cleanup, intricate parts handling, and laboratory analysis/technical work
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Thickness (mil)5
  • SizeLarge
  • Length (in)9 1/2
  • Length (cm)24
  • ColorBlue
  • GripTextured
  • ComplianceFDA-accepted
  • SterileNo
  • DescriptionBlue Nitrile Gloves, Powder Free, Large
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
TNT Blue disposable nitrile gloves are durable yet soft for improved comfort. They feature textured fingers for exceptional grip. Gloves are also static dissipative. Ambidextrous.