This item is now discontinued. Please browse similar items
Animal Temperature Probe for Mice, Type T; 1/16" Dia x 3/4" L, 5’ Cable
Representative image only.
Item # RK-92002-51
Accurate to 0.1°C within the physiological range
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeT
  • Sheath Diameter (in)0.0625
  • DescriptionAnimal Temperature Probe for Mice, Type T; 1/16\" Dia x 3/4\" L, 5’ Cable
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
These miniature Type T thermocouple sensors provide the widest and most versitile selection of temperature probes for bioscience and analytical chemistry applications. Models 08506-70 and 08506-75 (flexible implantable microprobes) are used for implantation in tissue, spectrophotometer cuvettes, water baths and more. Custom probes are available.