This item is now discontinued. Please browse similar items
Analtech TLC Developing Tank with Lid, Thinline™, Glass, Holds Two 10 x 10 cm Plates
Representative image only.
Analtech Mfr # A70-36 – Item # RK-34105-00
Designed for the development of TLC plates
  • Tanks are constructed of durable glass and racks are constructed of anodized aluminum
SPECS & DESCRIPTION
Specifications & Description
  • Backing MaterialGlass
  • Plate Length (cm)10
  • Plate Width (cm)10
  • Plate Capacity2
  • Descriptiondeveloping thinline tank with lid, capacity: two 10 x 10 cm plates
Key Features
  • Tanks are constructed of durable glass and racks are constructed of anodized aluminum
More About this Item
Tanks accommodate standard TLC plates. The top surface is ground to a precise tolerance for tight sealing to enable tank saturation and optimal plate development. Thinline™ tanks accommodate two plates and require less solvent for development. Latch-Lid™ units hold lids in place with stainless steel latches. Order rack to develop maximum number of plates in a tank.