This item is now discontinued. Please browse similar items
Amprobe Ultrasonic Gas Leak Detector
Representative image only.
Amprobe Mfr # ULD-300 – Item # RK-86417-00
“Listen” for pressurized gas leaks
  • Detects the ultrasonic noise created by pressure or vacuum leaks of any gas, regardless of type: CFC's, HFC's, nitrogen, CO2, or steam
  • Unaffected by contaminants or windy conditions
  • Identifies failing solenoids, valves, and bearings
  • Includes he
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Battery Type9 V battery
  • Battery LifeApproximately 40 hrs
  • DescriptionUltrasonic Gas Leak Detector
  • Warranty1 year
1 Year Warranty
Key Features
  • Detects the ultrasonic noise created by pressure or vacuum leaks of any gas, regardless of type: CFC's, HFC's, nitrogen, CO2, or steam
  • Unaffected by contaminants or windy conditions
  • Identifies failing solenoids, valves, and bearings
  • Includes he
  • 1 year warranty
More About this Item
Ultrasonic leak detectors have a decided advantage over conventional leak detectors—they "listen” for leaks and alert you to their presence. There is no gas sensor to become clogged or faulty, and no gas interferences to worry about. This leak detector locates any pressurized gas leak. It is excellent for use with boilers, furnaces, heat exchangers, and chillers. You can also use it to inspect bearings on pumps and machinery, or detect arcing electrical switchgear from a safe distance. Even windy outdoor conditions won’t affect its performance. The high-intensity LED indicator provides good visibility indoors or out. Use the ultrasonic transmitter (sold separately) with the leak detector to amplify leaks in insufficiently pressurized applications. The transmitter is useful in checking gaskets or panes of glass. Ultrasonic waves produce a different sound when a gasket or surface is damaged.