This item is now discontinued. Please browse similar items
Amprobe SV-U Carrying Case for Voltage and Continuity Testers
Representative image only.
Amprobe Mfr # SV-U – Item # RK-20008-77
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCarrying Case for Voltage and Continuity Testers
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
  • Extremely easy to use, no buttons, no setups
  • Automatically turns on and off
  • Automatically selects AC voltage, DC voltage or continuity
  • Indicates seven levels of AC or DC voltage
  • Visual continuity indication (LED)
  • Polarity indication
  • Continuity buzzer
  • Safety: CAT III 600V
SPECIFICATIONS
  • AC Voltage: 24/120/208/240/277/480/600
  • DC Voltage: 6/12/24/36/48/110/220
  • Continuity: 0-85 kOhms