Economical solution for locating underground energized and de-energized wires, cables, and pipes
Three testing modes for best accuracy – Passive power (50/60 Hz), passive radio (RF), active T-3500 signal generator with induction mode or directly connected to the cable with test leads or optional clamp
Active mode depth measurement up to 16 ft (4.9 m)
Semi-automatic gain control – fast signal location and control
33 kHz active frequency – best frequency for general locating
Full featured transmitter – quality locating capability
DescriptionPipe Transmitter, for use with Model AT-3500
Warranty1 Year
Includes
Receiver, transmitter, connection cable, ground stake, and ground cable.
Key Features
Three testing modes for best accuracy – Passive power (50/60 Hz), passive radio (RF), active T-3500 signal generator with induction mode or directly connected to the cable with test leads or optional clamp
Active mode depth measurement up to 16 ft (4.9 m)
Semi-automatic gain control – fast signal location and control
33 kHz active frequency – best frequency for general locating
Full featured transmitter – quality locating capability
Precise pinpointing of buried services and exceptionally fast sound and meter response with backlight multiple-segment bargraph digital display and audible indicators
Rugged durable construction, weatherproof and drop resistant up to 5 ft
Eliminates exploratory digging in parking lots or under floors for buried pipes and wires
SC-3500 Signal Clamp
Use to induce tracing signal to pipes, wires or cables
Clamps around the wire, does not require direct connection to the bare metal of wire
Accommodates cables, wires, and pipes up to 4 in. (10 cm) in diameter
MLS55-3 Sonde Pipe Transmitter
Use to pinpoint places in pipes, ducts, etc.
Can be inserted to the pipe/duct using standard fish tape or a duct rodder
Accommodates pipes over 2 in. (5 cm) in diameter
1 year warranty
More About this Item
As a single active frequency locating instrument, the AT-3500 employs the proven 33 kHz frequency for most locating applications. Additional functionality is found in the highly sensitive power and radio modes for line avoidance at utility construction sites. Depth measurement is taken with the press of a button in all modes of operation (active and passive). Unit incorporates a rugged housing, automatic digital display, and high quality features providing greater productivity at lower operational costs.