This item is now discontinued. Please browse similar items
Amprobe ACD 1000A Digital Clamp-On Multimeter
Representative image only.
Amprobe Mfr # ACD-6 PRO – Item # RK-20003-33
Available for sale in the United States and Canada only Rugged design is ideal for industrial environments
  • Auto and manual ranging
  • Relative zero mode
  • Auto power off
  • Data hold
  • Large, easy to read display
  • Diode test
  • Audible continuity
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • AC Max (amps)1000
  • Safety RatingCAT III 600V
  • Description1000A Digital Clamp-On Multimeter
  • Warranty1 Year
1 Year Warranty
Key Features
  • Auto and manual ranging
  • Relative zero mode
  • Auto power off
  • Data hold
  • Large, easy to read display
  • Diode test
  • Audible continuity